missing translation for 'onlineSavingsMsg'
Learn More

4-Iodopyrazole 98.0+%, TCI America™

Catalog No. I06925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06925G 5 g
I069225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06925G Supplier TCI America Supplier No. I06925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3469-69-0
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.975
MDL Number MFCD00005244
InChI Key LLNQWPTUJJYTTE-UHFFFAOYSA-N
Synonym 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295
PubChem CID 77022
ChEBI CHEBI:45260
IUPAC Name 4-iodo-1H-pyrazole
SMILES C1=C(C=NN1)I

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodopyrazole