missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isothiocyanate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 2059-76-9
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041091
InChI Key SNLGBRZZFRSXHA-UHFFFAOYSA-N
Synonym 4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
PubChem CID 74938
IUPAC Name 1-iodo-4-isothiocyanatobenzene
SMILES IC1=CC=C(C=C1)N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
S00350EA
View Documents
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2059-76-9
261.08
SNLGBRZZFRSXHA-UHFFFAOYSA-N
74938
IC1=CC=C(C=C1)N=C=S
C7H4INS
MFCD00041091
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
1-iodo-4-isothiocyanatobenzene

Specifications

2059-76-9
97%
IC6H4NCS
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
IC1=CC=C(C=C1)N=C=S
261.08
261.08
4-Iodophenyl isothiocyanate
97.0
C7H4INS
MFCD00041091
SNLGBRZZFRSXHA-UHFFFAOYSA-N
1-iodo-4-isothiocyanatobenzene
74938
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents