missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iododibenzothiophene 98.0+%, TCI America™
$76.43 - $272.16
Chemical Identifiers
| CAS | 132034-89-0 |
|---|---|
| Molecular Formula | C12H7IS |
| Molecular Weight (g/mol) | 310.15 |
| MDL Number | MFCD00159895 |
| InChI Key | SKILYZCQRUBEIH-UHFFFAOYSA-N |
| PubChem CID | 458353 |
| IUPAC Name | 6-iodo-8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaene |
| SMILES | IC1=CC=CC2=C1SC1=C2C=CC=C1 |
Chemical Identifiers
| 132034-89-0 | |
| 310.15 | |
| SKILYZCQRUBEIH-UHFFFAOYSA-N | |
| 6-iodo-8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaene |
| C12H7IS | |
| MFCD00159895 | |
| 458353 | |
| IC1=CC=CC2=C1SC1=C2C=CC=C1 |
Specifications
| 132034-89-0 | |
| White-Yellow | |
| MFCD00159895 | |
| SKILYZCQRUBEIH-UHFFFAOYSA-N | |
| 6-iodo-8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaene | |
| 458353 | |
| ≥98.0% (GC) | |
| 4-Iododibenzothiophene |
| 102°C | |
| C12H7IS | |
| 1 g | |
| IC1=CC=CC2=C1SC1=C2C=CC=C1 | |
| 310.15 | |
| 310.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No