missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iododibenzofuran 98.0+%, TCI America™
$257.97 - $772.00
Chemical Identifiers
| CAS | 65344-26-5 |
|---|---|
| Molecular Formula | C12H7IO |
| Molecular Weight (g/mol) | 294.091 |
| MDL Number | MFCD00094364 |
| InChI Key | ACEYZYYNAMWIIE-UHFFFAOYSA-N |
| PubChem CID | 458257 |
| IUPAC Name | 4-iododibenzofuran |
| SMILES | C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I0972200MG
|
TCI America
I0972200MG |
200 mg |
Each for $257.97
|
|
|||||
|
I09721G
|
TCI America
I09721G |
1 g |
Each for $772.00
|
|
|||||
Chemical Identifiers
| 65344-26-5 | |
| 294.091 | |
| ACEYZYYNAMWIIE-UHFFFAOYSA-N | |
| 4-iododibenzofuran |
| C12H7IO | |
| MFCD00094364 | |
| 458257 | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)I |
Specifications
| 65344-26-5 | |
| White-Yellow | |
| MFCD00094364 | |
| ACEYZYYNAMWIIE-UHFFFAOYSA-N | |
| 4-iododibenzofuran | |
| 458257 | |
| ≥98.0% (GC) | |
| 4-Iododibenzofuran |
| 73°C | |
| C12H7IO | |
| 200 mg | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)I | |
| 294.091 | |
| 294.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No