missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobutyronitrile, 97%

Catalog No. AC390390010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390390010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC390390010 Supplier Thermo Scientific Chemicals Supplier No. 390390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6727-73-7 | C4H6IN | 195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6727-73-7
Molecular Formula C4H6IN
Molecular Weight (g/mol) 195
MDL Number MFCD00039501
InChI Key CWINCVBWHUGBEB-UHFFFAOYSA-N
Synonym butanenitrile, 4-iodo, g-iodobutyronitrile, 4-iodobutyronitrile
PubChem CID 285648
IUPAC Name 4-iodobutanenitrile
SMILES C(CC#N)CI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.8260g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.5370
Linear Formula I(CH2)3CN
Quantity 1 g
Specific Gravity 1.826
Formula Weight 195
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Iodobutyronitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only