missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobutyl acetate, 96%, stab. with copper

Catalog No. AAH3182618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3182618 50 g
AAH3182609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3182618 Supplier Thermo Scientific Chemicals Supplier No. H3182618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40596-44-9 | C6H11IO2 | 242.056 g/mol

4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobutyl acetate is used in the preparation of tricyclohexyl(acetoxybutyl))phosphonium iodide, tri-n-hexyl(acetoxybutyl))phosphonium iodide

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 40596-44-9
Molecular Formula C6H11IO2
Molecular Weight (g/mol) 242.056
MDL Number MFCD00010664
InChI Key FLPVVTDEKLGZDZ-UHFFFAOYSA-N
Synonym tech., 4-iodobutylacetate, 4-acetoxybutyl iodide, 4-iodo-butyl acetate, acmc-20amg9, acetic acid 4-iodobutyl ester, acetic acid 4-iodo-butyl ester, 1-butanol, 4-iodo-,1-acetate, 4-iodobutyl acetate, technical grade
PubChem CID 2733405
IUPAC Name 4-iodobutyl acetate
SMILES CC(=O)OCCCCI

Specifications

Density 1.61
Boiling Point 94°C to 95°C (5.5 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.5115
Linear Formula CH3CO2(CH2)4I
Quantity 50 g
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 242.05
Percent Purity 96%
Chemical Name or Material 4-Iodobutyl acetate, stabilized with copper
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only