missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AC412390050
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC412390050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412390050 Supplier Thermo Scientific Chemicals Supplier No. 412390050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1711-02-0
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H4ClIO
Linear Formula IC6H4COCl
Quantity 5g
MDL Number MFCD00001058
Beilstein 09, 367
Synonym p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
InChI Key NJAKCIUOTIPYED-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=O)Cl)I
IUPAC Name 4-iodobenzoyl chloride
Molecular Weight (g/mol) 266.46
PubChem CID 74373
Formula Weight 266.46
Percent Purity 98%
Chemical Name or Material 4-Iodobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-974-7
TSCA TSCA

RUO – Research Use Only