missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzenesulfonyl Chloride 93.0+%, TCI America™

Catalog No. p-7131180
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00515G 5 g
I005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00515G Supplier TCI America Supplier No. I00515G

Chemical Identifiers

CAS 98-61-3
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00007441
InChI Key POXFXTSTVWDWIR-UHFFFAOYSA-N
Synonym pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog
PubChem CID 7399
IUPAC Name 4-iodobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)I

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 302.51
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzenesulfonyl Chloride