Learn More
4-Iodobenzenesulfonamide, 95%, Thermo Scientific™
$79.53 - $201.41
Chemical Identifiers
| CAS | 825-86-5 |
|---|---|
| Molecular Formula | C6H6INO2S |
| Molecular Weight (g/mol) | 283.09 |
| InChI Key | KQZFABSTXSNEQH-UHFFFAOYSA-N |
| Synonym | p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference |
| PubChem CID | 13218 |
| IUPAC Name | 4-iodobenzenesulfonamide |
| SMILES | C1=CC(=CC=C1S(=O)(=O)N)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC455700010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC455700050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 825-86-5 | |
| 283.09 | |
| p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference | |
| 4-iodobenzenesulfonamide |
| C6H6INO2S | |
| KQZFABSTXSNEQH-UHFFFAOYSA-N | |
| 13218 | |
| C1=CC(=CC=C1S(=O)(=O)N)I |
Specifications
| 825-86-5 | |
| Authentic | |
| Glass Bottle | |
| IC6H4SO2NH2 | |
| KQZFABSTXSNEQH-UHFFFAOYSA-N | |
| 4-iodobenzenesulfonamide | |
| 13218 | |
| 95% |
| 1% max. | |
| 94% min. (HPLC) | |
| C6H6INO2S | |
| p-iodobenzenesulfonamide, 4-iodobenzenesulphonamide, 4-iodobenzene-1-sulfonamide, benzenesulfonamide,4-iodo, benzenesulfonamide, p-iodo, p-jodbenzolsulfonamid, 4-iodibenzenesulphonamide, 4-iodo-benzenesulfonamide, 4-iodo-1-sulfamoylbenzene, 3-11-00-00110 beilstein handbook reference | |
| C1=CC(=CC=C1S(=O)(=O)N)I | |
| 283.09 | |
| 283.09 | |
| 4-Iodobenzenesulfonamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only