missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone 98.0+%, TCI America™

Catalog No. p-7131321
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I03055G 5 g
I030525G 25 g
2 options

Catalog No. I03055G

Supplier: TCI America I03055G

Only null left

Chemical Identifiers

CAS 13329-40-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00045320
InChI Key JZJWCDQGIPQBAO-UHFFFAOYSA-N
Synonym 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
PubChem CID 72869
IUPAC Name 1-(4-iodophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)I

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 246.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Iodoacetophenone