missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone, 98%

CAS: 13329-40-3 | C8H7IO | 246.047 g/mol

Supplier:  Thermo Scientific Chemicals A1679406

 View more versions of this product

Catalog No. AAA1679406


Only null left
Explore available promotions

Description

Description

Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4'-Iodoacetophenone
82°C to 84°C
MFCD00045320
1857412
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
JZJWCDQGIPQBAO-UHFFFAOYSA-N
1-(4-iodophenyl)ethanone
72869
98%
13329-40-3
C8H7IO
5 g
Light sensitive
Easily soluble in water.
CC(=O)C1=CC=C(C=C1)I
246.047
246.05

Chemical Identifiers

13329-40-3
246.047
JZJWCDQGIPQBAO-UHFFFAOYSA-N
72869
CC(=O)C1=CC=C(C=C1)I
C8H7IO
MFCD00045320
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
1-(4-iodophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-372-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only