missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-L-phenylalanine, 95%

Catalog No. p-7034308
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197903 1 g
AAH5197906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197903 Supplier Thermo Scientific Chemicals Supplier No. H5197903

CAS: 24250-85-9 | C9H10INO2 | 291.09 g/mol

4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

Solubility
Partly miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 24250-85-9
Molecular Formula C9H10INO2
Molecular Weight (g/mol) 291.09
MDL Number MFCD00002602
InChI Key PZNQZSRPDOEBMS-QMMMGPOBSA-N
Synonym 4-iodo-l-phenylalanine, 4-iodophenylalanine, l-4-iodophenylalanine, iodo-phenylalanine, p-iodo-l-phenylalanine, p-iodophenylalanine, s-2-amino-3-4-iodophenyl propanoic acid, 2s-2-amino-3-4-iodophenyl propanoic acid, h-phe 4-i-oh, phenylalanine, 4-iodo
PubChem CID 134497
IUPAC Name (2S)-2-amino-3-(4-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(I)C=C1)C(O)=O

Specifications

Beilstein 4411317
Solubility Information Partly miscible with water.
Quantity 1 g
Formula Weight 291.09
Optical Rotation −6.5° (c=1 in Acetic Acid:Water (4:1))
Percent Purity 95%
Sensitivity Light sensitive
Chemical Name or Material 4-Iodo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only