missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-4',4″-dimethoxytriphenylamine 98.0+%, TCI America™

Catalog No. I07761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07761G Supplier TCI America Supplier No. I07761G
Only null left

Chemical Identifiers

CAS 201802-15-5
Molecular Formula C20H18INO2
Molecular Weight (g/mol) 431.273
InChI Key VGQDQJMYUYSINZ-UHFFFAOYSA-N
Synonym 4-Iodo-N,N-bis(4-methoxyphenyl)aniline
PubChem CID 44630018
IUPAC Name N-(4-iodophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)I

Specifications

Melting Point 111°C
Quantity 1 g
Formula Weight 431.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-4′,4′′-dimethoxytriphenylamine