missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea, 95%

Catalog No. AC393740050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393740050 5 g Glass Bottle
AC393740250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393740050 Supplier Thermo Scientific Chemicals Supplier No. 393740050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107819-90-9 | C12H22N2O4S | 290.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107819-90-9
Molecular Formula C12H22N2O4S
Molecular Weight (g/mol) 290.38
MDL Number MFCD00239356
InChI Key UQJXXWHAJKRDKY-UHFFFAOYSA-N
Synonym 1,3-di-boc-2-methylisothiourea, s-methyl-n,n'-bis tert-butoxycarbonyl isothiourea, 1,3-bis tert-butoxycarbonyl-2-methyl-2-thiopseudourea, n,n'-di-boc-s-methylisothiourea, n,n inverted exclamation marka-di-boc-s-methylisothiourea, n,n-di-boc-s-methylisothiourea, bis-boc-thiopseudourea, polymer-bound, 1,3-di-tert-butoxycarbonyl-2-methylisothiourea, 1,3-bis-tert-butoxycarbonyl-2-methyl-thiopseudourea, z-n,n'-bis tert-butyloxycarbonyl-s-methylisothiourea
PubChem CID 6376008
IUPAC Name tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-methylsulfanylmethylidene]carbamate
SMILES CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)SC

Specifications

Melting Point 115.0°C to 121.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 290.38
Percent Purity 95%
Chemical Name or Material 1, 3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only