missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™

Catalog No. p-2701853
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09111CB 250mg
CC09111DA 1g
CC09111DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC09111CB Supplier Thermo Scientific Chemicals Supplier No. CC09111CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 352018-95-2
Molecular Formula C6H3IN2S
Molecular Weight (g/mol) 262.07
MDL Number MFCD02681904
InChI Key ROPRWVSWECJQIC-UHFFFAOYSA-N
Synonym 4-iodobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,4-iodo, 4-iodo-benzo 1,2,5 thiadiazole, 4-iodanyl-2,1,3-benzothiadiazole, 4-iodobenzo c 1,2,5-thiadiazole, 4-iodo-2,1,3-benzothiadiazole
PubChem CID 2776287
IUPAC Name 4-iodo-2,1,3-benzothiadiazole
SMILES IC1=CC=CC2=NSN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 262.07
Percent Purity 97%
Chemical Name or Material 4-Iodo-2, 1, 3-benzothiadiazole
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.