missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1H-imidazole 98.0+%, TCI America™

Catalog No. I08125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08125G 5 g
I08121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08125G Supplier TCI America Supplier No. I08125G

Chemical Identifiers

CAS 71759-89-2
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.98
MDL Number MFCD01632213
InChI Key BHCMXJKPZOPRNN-UHFFFAOYSA-N
Synonym 4-iodoimidazole, 4-iodo-1h-imidazole, 4 5-iodoimidazole, 4 5-iodo-1 h-imidazole, 4-iodo-1 h-imidazole, 4-iodo-3h-imidazole, 1h-imidazole, 4-iodo, 5-iodoimidazole, 4-iodo-imidazole, sftheqvilimkp@
PubChem CID 606522
IUPAC Name 5-iodo-1H-imidazole
SMILES IC1=CN=CN1

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-1H-imidazole