missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-1-methylpyrazole 98.0+%, TCI America™
$78.29 - $233.00
Chemical Identifiers
| CAS | 39806-90-1 |
|---|---|
| Molecular Formula | C4H5IN2 |
| Molecular Weight (g/mol) | 208.002 |
| MDL Number | MFCD05663860 |
| InChI Key | RSDRDHPLXWMTRJ-UHFFFAOYSA-N |
| Synonym | 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole |
| PubChem CID | 2734773 |
| IUPAC Name | 4-iodo-1-methylpyrazole |
| SMILES | CN1C=C(C=N1)I |
Chemical Identifiers
| 39806-90-1 | |
| 208.002 | |
| RSDRDHPLXWMTRJ-UHFFFAOYSA-N | |
| 2734773 | |
| CN1C=C(C=N1)I |
| C4H5IN2 | |
| MFCD05663860 | |
| 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole | |
| 4-iodo-1-methylpyrazole |
Specifications
| 39806-90-1 | |
| White | |
| MFCD05663860 | |
| 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole | |
| CN1C=C(C=N1)I | |
| 208.002 | |
| 208.00 | |
| Crystalline Powder |
| 64°C | |
| C4H5IN2 | |
| 1 g | |
| RSDRDHPLXWMTRJ-UHFFFAOYSA-N | |
| 4-iodo-1-methylpyrazole | |
| 2734773 | |
| ≥98.0% (GC) | |
| 4-Iodo-1-methylpyrazole |
Safety and Handling
TSCA : No