missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-methylpyrazole 98.0+%, TCI America™

Catalog No. I08291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08291G 1 g
I08295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08291G Supplier TCI America Supplier No. I08291G

Chemical Identifiers

CAS 39806-90-1
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD05663860
InChI Key RSDRDHPLXWMTRJ-UHFFFAOYSA-N
Synonym 4-iodo-1-methyl-1h-pyrazole, 1-methyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-methyl, 1-methyl-4-iodopyrazole, zlchem 32, pubchem24346, acmc-1ctkh, n-methyl-4-iodopyrazole, 1-methyl-4-iodo pyrazole, 4-iodo-1-methyl-pyrazole
PubChem CID 2734773
IUPAC Name 4-iodo-1-methylpyrazole
SMILES CN1C=C(C=N1)I

Specifications

Melting Point 64°C
Color White
Quantity 1 g
Formula Weight 208.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-1-methylpyrazole