missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-methylpyrazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. I07861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07861G 1 g
I07865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07861G Supplier TCI America Supplier No. I07861G

Chemical Identifiers

CAS 75092-30-7
Molecular Formula C5H5IN2O2
Molecular Weight (g/mol) 252.011
MDL Number MFCD00461121
InChI Key SZRMOKQNOJYMNK-UHFFFAOYSA-N
PubChem CID 673687
IUPAC Name 4-iodo-2-methylpyrazole-3-carboxylic acid
SMILES CN1C(=C(C=N1)I)C(=O)O

Specifications

Melting Point 242°C
Color White
Quantity 1 g
Formula Weight 252.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-1-methylpyrazole-5-carboxylic Acid