missing translation for 'onlineSavingsMsg'
Learn More

4-Imidazolecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7131512
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06071G 1 g
I06075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06071G Supplier TCI America Supplier No. I06071G
Only null left

Chemical Identifiers

CAS 1072-84-0
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00082203
InChI Key NKWCGTOZTHZDHB-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carboxylic acid, 4-imidazolecarboxylic acid, imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4-carboxyimidazole, unii-ae82z8u4gj, imidazole-5-carboxylic acid, 4-imidazole carboxylic acid, ae82z8u4gj, 4 5-imidazolecarboxylic acid
PubChem CID 14080
IUPAC Name 1H-imidazole-5-carboxylic acid
SMILES OC(=O)C1=CN=CN1

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 1 g
Formula Weight 112.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Imidazolecarboxylic Acid
RTECSNumber NI4001000
TSCA No
Recommended Storage Refrigerator