missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyvalerophenone 98.0+%, TCI America™

Catalog No. p-7130644
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
H062725G 25 g
H0627500G 500 g
2 options

Catalog No. H062725G

Supplier: TCI America H062725G

Only null left

Chemical Identifiers

CAS 2589-71-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009719
InChI Key ZKCJJGOOPOIZTE-UHFFFAOYSA-N
Synonym 4'-hydroxyvalerophenone, 4-valerylphenol, 1-pentanone, 1-4-hydroxyphenyl, p-valerylphenol, 1-4-hydroxyphenyl pentan-1-one, p-hydroxyvalerophenone, 4'-hydroxypentanophenone, valerophenone, 4'-hydroxy, 4-pentanoylphenol
PubChem CID 75766
IUPAC Name 1-(4-hydroxyphenyl)pentan-1-one
SMILES CCCCC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyvalerophenone