missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxythiobenzamide 98.0+%, TCI America™

Catalog No. H13085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13085G 5 g
H130825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13085G Supplier TCI America Supplier No. H13085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25984-63-8
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973332
InChI Key DOCQBKMMNPJLOR-UHFFFAOYSA-N
Synonym 4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
PubChem CID 5706487
IUPAC Name 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES NC(S)=C1C=CC(=O)C=C1

Specifications

Melting Point 192°C
Color Yellow
Quantity 5 g
Formula Weight 153.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxythiobenzamide