missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxythiobenzamide, 98%

Catalog No. AAH3278014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3278014 25 g
AAH3278006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3278014 Supplier Thermo Scientific Chemicals Supplier No. H3278014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25984-63-8 | C7H7NOS | 153.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25984-63-8
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973332
InChI Key DOCQBKMMNPJLOR-UHFFFAOYSA-N
Synonym 4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
PubChem CID 5706487
IUPAC Name 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES NC(S)=C1C=CC(=O)C=C1

Specifications

Melting Point 181°C to 185°C
Quantity 25 g
Formula Weight 153.2
Percent Purity 98%
Chemical Name or Material 4-Hydroxythiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only