missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

Catalog No. p-7077151
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC121910050 5 g
AC121910250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC121910050 Supplier Thermo Scientific Chemicals Supplier No. 121910050
Only null left

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 345909-35-5
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00149476
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
Synonym 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
PubChem CID 18530299
IUPAC Name 4-oxo-1H-quinoline-2-carboxylic acid;hydrate
SMILES OC(=O)C1=NC2=CC=CC=C2C(O)=C1

Specifications

Melting Point 275.0°C
Infrared Spectrum Authentic
Quantity 5 g
Beilstein 22,23
Merck Index 14,5327
Solubility Information Solubility in water: 0.9g/L water (100°C). Other solubilities: soluble in hot alcohol, insoluble in ether, very soluble in alkali
Formula Weight 189.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid, hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only