Learn More
4-Hydroxyquinoline-2-carboxylic acid hydrate, 98%
CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol
Supplier: Thermo Scientific Chemicals A1260214
Description
Broad spectrum excitatory amino acid antagonist4-Hydroxyquinoline-2-carboxylic acid hydrate is used as an intermediate in synthetic chemistry. It acts as an antiexcitotoxic and anticonvulsant. It finds application in certain neurobiological disorders therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in hot ethanol and water. Insoluble in ether.Â
Notes
Incompatible with oxidizing agents.
Specifications
| 4-Hydroxyquinoline-2-carboxylic acid hydrate | |
| ∼275°C (decomposition) | |
| MFCD00149476 | |
| 147451 | |
| 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1 | |
| HCZHHEIFKROPDY-UHFFFAOYSA-N | |
| 4-oxo-1H-quinoline-2-carboxylic acid;hydrate | |
| 18530299 | |
| 98% |
| 345909-35-5 | |
| C10H7NO3 | |
| 25 g | |
| 14,5327 | |
| Soluble in hot ethanol and water. Insoluble in ether. | |
| OC(=O)C1=NC2=CC=CC=C2C(O)=C1 | |
| 189.17 | |
| 189.17 (Anhydrous) |
Chemical Identifiers
| 345909-35-5 | |
| 189.17 | |
| HCZHHEIFKROPDY-UHFFFAOYSA-N | |
| 18530299 |
| C10H7NO3 | |
| MFCD00149476 | |
| 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1 | |
| OC(=O)C1=NC2=CC=CC=C2C(O)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-751-5
RTECSNumber : VB2080000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only