missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinazoline, 98%

Catalog No. AAA1712918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1712918 50 g
AAA1712909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1712918 Supplier Thermo Scientific Chemicals Supplier No. A1712918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 491-36-1 | C8H6N2O | 146.149 g/mol

4-Hydroxyquinazoline is been used to inhibit PARP (poly(ADP-ribose) synthetase) which catalyzes covalent attachment of the ADP-ribose moiety of NAD+ to various proteins. This compound specifically and potently inhibits PARP-1. 4-HQN demonstrates the ability to decrease activation of transcription factor NFκB and AP-1 in lipopolysaccharide-induced shock. Mechanistic studies indicate that 4-HQN activates PI3-kinase/Akt pathway in the liver, spleen, and lung and down-regulates two elements of the MAP kinase system. Additionally, this agent has been observed to decrease ischemia-reperfusion-induced increase of protein oxidation, single-strand DNA breaks, lipid peroxidation, and mitochondrial reactive oxygen species production in the reperfusion period.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyquinazoline is been used to inhibit PARP (poly(ADP-ribose) synthetase) which catalyzes covalent attachment of the ADP-ribose moiety of NAD+ to various proteins. This compound specifically and potently inhibits PARP-1. 4-HQN demonstrates the ability to decrease activation of transcription factor NFκB and AP-1 in lipopolysaccharide-induced shock. Mechanistic studies indicate that 4-HQN activates PI3-kinase/Akt pathway in the liver, spleen, and lung and down-regulates two elements of the MAP kinase system. Additionally, this agent has been observed to decrease ischemia-reperfusion-induced increase of protein oxidation, single-strand DNA breaks, lipid peroxidation, and mitochondrial reactive oxygen species production in the reperfusion period.

Solubility
Soluble in DMSO (100 mM), ethanol, methanol, and water (10 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 491-36-1
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00511302
InChI Key QMNUDYFKZYBWQX-UHFFFAOYSA-N
Synonym 4-hydroxyquinazoline, 4-quinazolinol, quinazolin-4-ol, quinazolin-4 3h-one, 4 3h-quinazolinone, 4-quinazolone, 4-quinazolinone, 4 1h-quinazolinone, 4-oxoquinazoline, 3,4-dihydroquinazolin-4-one
PubChem CID 63112
IUPAC Name 1H-quinazolin-4-one
SMILES C1=CC=C2C(=C1)C(=O)N=CN2

Specifications

Melting Point 216°C to 219°C
Odor Odorless
Quantity 50 g
Beilstein 118473
Solubility Information Soluble in DMSO (100 mM),ethanol,methanol,and water (10 mM).
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinazoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-735-8
RTECSNumber VA2300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only