missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypyridine, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Melting Point | 140.0°C to 147.0°C |
| Color | Beige to Brown |
| Density | 221g/mL |
| Boiling Point | 230.0°C to 235.0°C (12.0 mmHg) |
| Flash Point | 221°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 210-958-3 |
RUO – Research Use Only