missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypiperidine 98.0+%, TCI America™

$90.48 - $254.11

Chemical Identifiers

CAS 5382-16-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00005999
InChI Key HDOWRFHMPULYOA-UHFFFAOYSA-N
Synonym 4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
PubChem CID 79341
IUPAC Name piperidin-4-ol
SMILES C1CNCCC1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H041325G
View Documents
TCI America
H041325G
25 g
Each for $90.48
Only null left
 
H0413100G
View Documents
TCI America
H0413100G
100 g
Each for $254.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5382-16-1
101.149
HDOWRFHMPULYOA-UHFFFAOYSA-N
79341
C1CNCCC1O
C5H11NO
MFCD00005999
4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
piperidin-4-ol

Specifications

5382-16-1
White-Yellow
C5H11NO
25 g
HDOWRFHMPULYOA-UHFFFAOYSA-N
piperidin-4-ol
79341
≥98.0% (GC,T)
4-Hydroxypiperidine
88°C
114°C
MFCD00005999
4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
C1CNCCC1O
101.149
101.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2218

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'