missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypiperidine 98.0+%, TCI America™

Catalog No. H041325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H041325G 25 g
H0413100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H041325G Supplier TCI America Supplier No. H041325G

Chemical Identifiers

CAS 5382-16-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00005999
InChI Key HDOWRFHMPULYOA-UHFFFAOYSA-N
Synonym 4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
PubChem CID 79341
IUPAC Name piperidin-4-ol
SMILES C1CNCCC1O

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 114°C
Quantity 25 g
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxypiperidine
EINECSNumber (9)-2218
TSCA No