missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypiperidine Hydrochloride 98.0+%, TCI America™

Catalog No. h068625g
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
H068625G 25 g
1 options

Catalog No. H068625G

Supplier: TCI America H068625G

Only null left

Chemical Identifiers

CAS 5382-17-2
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD00035298
InChI Key VKCORPXOKYDINR-UHFFFAOYSA-N
Synonym 4-Piperidinol Hydrochloride
PubChem CID 16212499
IUPAC Name piperidin-4-ol;hydrochloride
SMILES C1CNCCC1O.Cl

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 25 g
Formula Weight 137.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxypiperidine Hydrochloride
EINECSNumber (9)-2218
TSCA No