missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethylbenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 394340100

Quantity 10g
Packaging Glass Bottle

 View more versions of this product

Catalog No. AC394340100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3006-96-0
152.15
WWYFPDXEIFBNKE-UHFFFAOYSA-N
76360
OCC1=CC=C(C=C1)C(O)=O
C8H8O3
MFCD00017598
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
4-(hydroxymethyl)benzoic acid

Specifications

4-Hydroxymethylbenzoic acid
97.5
Authentic
Glass Bottle
COOHC6H4CH2OH
10g
WWYFPDXEIFBNKE-UHFFFAOYSA-N
4-(hydroxymethyl)benzoic acid
76360
98%
3006-96-0
100.0
97.5% min. (HPLC)
C8H8O3
MFCD00017598
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
OCC1=CC=C(C=C1)C(O)=O
152.15
152.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents