Learn More
4-(Hydroxymethyl)tetrahydropyran, 98%
CAS: 14774-37-9 | C6H12O2 | 116.16 g/mol
Supplier: Thermo Scientific Chemicals H6456803
Description
Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 4-(Hydroxymethyl)tetrahydropyran | |
| 111°C to 115°C (15 mmHg) | |
| C6H12O2 | |
| MFCD00457804 | |
| 103069 | |
| Slightly soluble in water. | |
| OCC1CCOCC1 | |
| 116.16 | |
| 116.16 |
| 14774-37-9 | |
| 61°C (142°F) | |
| 1.46 | |
| 1 g | |
| tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
| YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
| oxan-4-ylmethanol | |
| 2773573 | |
| 98% |
Chemical Identifiers
| 14774-37-9 | |
| 116.16 | |
| YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
| 2773573 |
| C6H12O2 | |
| MFCD00457804 | |
| tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
| OCC1CCOCC1 |
Safety and Handling
GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only