missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Hydroxymethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
$316.89 - $1,066.24
Chemical Identifiers
| CAS | 13380-84-2 |
|---|---|
| Molecular Formula | C8H14O3 |
| Molecular Weight (g/mol) | 158.20 |
| MDL Number | MFCD10566907,MFCD01734677,MFCD10566906 |
| InChI Key | VQMIUUBKKPIDBN-UHFFFAOYSA-N |
| Synonym | 4-Carboxy-1-cyclohexanemethanol |
| PubChem CID | 202819 |
| IUPAC Name | 4-(hydroxymethyl)cyclohexane-1-carboxylic acid |
| SMILES | OCC1CCC(CC1)C(O)=O |
Chemical Identifiers
| 13380-84-2 | |
| 158.20 | |
| VQMIUUBKKPIDBN-UHFFFAOYSA-N | |
| 202819 | |
| OCC1CCC(CC1)C(O)=O |
| C8H14O3 | |
| MFCD10566907,MFCD01734677,MFCD10566906 | |
| 4-Carboxy-1-cyclohexanemethanol | |
| 4-(hydroxymethyl)cyclohexane-1-carboxylic acid |
Specifications
| 13380-84-2 | |
| C8H14O3 | |
| 5 g | |
| VQMIUUBKKPIDBN-UHFFFAOYSA-N | |
| 4-(hydroxymethyl)cyclohexane-1-carboxylic acid | |
| 202819 | |
| ≥98.0% (GC,T) | |
| 4-(Hydroxymethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) |
| Red-White | |
| MFCD10566907,MFCD01734677,MFCD10566906 | |
| 4-Carboxy-1-cyclohexanemethanol | |
| OCC1CCC(CC1)C(O)=O | |
| 158.20 | |
| 158.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GU8582500
TSCA : No