missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl-5-methylimidazole Hydrochloride 98.0+%, TCI America™

Catalog No. H062525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H062525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H062525G Supplier TCI America Supplier No. H062525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38585-62-5
Molecular Formula C5H9ClN2O
Molecular Weight (g/mol) 148.59
MDL Number MFCD00040986
InChI Key UBHDUFNPQJWPRQ-UHFFFAOYSA-N
Synonym 4-methyl-5-imidazolemethanol hydrochloride, 4-methyl-1h-imidazol-5-yl methanol hydrochloride, 5-methyl-1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethyl-5-methylimidazole hydrochloride, 4-methylimidazol-5-ylmethanolhydrochloride, 4-methyl-5-hydroxymethyl imidazole hydrochloride, 1h-imidazole-4-methanol, 5-methyl-, monohydrochloride, 5-hydroxymethyl-4-methylimidazole hcl, 4-methyl-5-hydroxymethylimidazole hydrochloride, 4-methyl-5-hydroxymethyl-imidazole hydrochloride
PubChem CID 3084707
IUPAC Name hydrogen (5-methyl-1H-imidazol-4-yl)methanol chloride
SMILES [H+].[Cl-].CC1=C(CO)N=CN1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 148.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxymethyl-5-methylimidazole Hydrochloride