missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyisophthalic Acid 98.0+%, TCI America™

Catalog No. H025725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H025725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H025725G Supplier TCI America Supplier No. H025725G

Chemical Identifiers

CAS 636-46-4
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00010391
InChI Key BCEQKAQCUWUNML-UHFFFAOYSA-N
Synonym 4-hydroxyisophthalic acid, eupirina, 4-hydroxy-m-phthalic acid, isophthalic acid, 4-hydroxy, 4-hipa, 4-hydroxy-1,3-benzenedicarboxylic acid, unii-p87ff77o00, 1,3-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxy isophthalic acid, 4-hydroxyisophthalate
PubChem CID 12490
IUPAC Name 4-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 182.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyisophthalic Acid
EINECSNumber (9)-1095
RTECSNumber NT2544000
TSCA No