missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxyisophthalic Acid, 98%, Spectrum™ Chemical
Description
Spectrum™ Chemical 4-Hydroxyisophthalic Acid
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Percent Purity | 98% |
| Grade | Reagent |