missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyisophthalic Acid, 98%, Spectrum™ Chemical

Catalog No. 18600140
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600140 1 kg Amber Glass Bottle
18600138 5 g Amber Glass Bottle
18600139 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600140 Supplier Spectrum Chemical Mfg Cor Supplier No. H23701KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

H2370, 636-46-4, C8H6O5

Spectrum™ Chemical 4-Hydroxyisophthalic Acid

Chemical Identifiers

CAS 636-46-4
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.13
InChI Key BCEQKAQCUWUNML-UHFFFAOYSA-N
IUPAC Name 4-hydroxybenzene-1,3-dicarboxylic acid
SMILES OC(=O)C1=CC=C(O)C(=C1)C(O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98%
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material 4-Hydroxyisophthalic Acid