missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyisophthalaldehyde 98.0+%, TCI America™

Catalog No. p-7129608
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03101G 1 g
F03105G 5 g
F031025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F03101G Supplier TCI America Supplier No. F03101G
Only null left

Chemical Identifiers

CAS 3328-70-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00003334
InChI Key FEUATHOQKVGPEK-UHFFFAOYSA-N
Synonym 2,4-Diformylphenol
PubChem CID 165106
IUPAC Name 4-hydroxybenzene-1,3-dicarbaldehyde
SMILES OC1=CC=C(C=O)C=C1C=O

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 1 g
Formula Weight 150.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyisophthalaldehyde