missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyindole, 98%

Catalog No. AAL1435203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1435203 1 g
AAL1435206 5 g
AAL1435214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1435203 Supplier Thermo Scientific Chemicals Supplier No. L1435203

CAS: 2380-94-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2380-94-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005667
InChI Key NLMQHXUGJIAKTH-UHFFFAOYSA-N
Synonym 4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
PubChem CID 75421
ChEBI CHEBI:24702
IUPAC Name 1H-indol-4-ol
SMILES OC1=C2C=CNC2=CC=C1

Specifications

Melting Point 97°C to 99°C
Quantity 1 g
Beilstein 114905
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxyindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-177-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only