missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyheptanophenone 98.0+%, TCI America™

Catalog No. p-7130770
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H086425G 25 g
H0864500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H086425G Supplier TCI America Supplier No. H086425G

Chemical Identifiers

CAS 14392-72-4
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00191523
InChI Key MAZPQHJYGKZQEA-UHFFFAOYSA-N
Synonym Hexyl 4-Hydroxyphenyl Ketone
PubChem CID 578426
IUPAC Name 1-(4-hydroxyphenyl)heptan-1-one
SMILES CCCCCCC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 25 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyheptanophenone