missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin 98.0+%, TCI America™

Catalog No. p-7130428
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023525G 25 g
H0235250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H023525G Supplier TCI America Supplier No. H023525G
Only null left

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key OWBBAPRUYLEWRR-UHFFFAOYSA-N
Synonym 4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
IUPAC Name 2-hydroxy-4H-chromen-4-one
SMILES OC1=CC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 212°C
Color Yellow
Quantity 25 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxycoumarin
RTECSNumber DJ3100000
TSCA Yes