missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin, 98%

Catalog No. AC121100050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121100050 5 g Glass bottle
AC121101000 100 g Plastic bottle
AC121105000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121100050 Supplier Thermo Scientific Chemicals Supplier No. 121100050

CAS: 1076-38-6 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key OWBBAPRUYLEWRR-UHFFFAOYSA-N
Synonym 4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
SMILES OC1=CC(=O)C2=CC=CC=C2O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Colorless to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 488
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in hot water,soluble in alcohol,acetone and ether
IUPAC Name 4-hydroxychromen-2-one
Formula Weight 162.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Hydroxycoumarin, 98%
Show More Show Less
EINECSNumber 214-060-2
RTECSNumber DJ3100000
TSCA TSCA

RUO – Research Use Only