missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybutyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 95°C |
| Quantity | 100 g |
| Formula Weight | 144.17 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Hydroxybutyl Acrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-0958 |
| TSCA | Yes |