missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybutyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. A1390500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1390500G 500 g
A139025G 25 g
A1390100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1390500G Supplier TCI America Supplier No. A1390500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2478-10-6
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00010261
InChI Key NDWUBGAGUCISDV-UHFFFAOYSA-N
Synonym 4-hydroxybutyl acrylate, acrylic acid 4-hydroxybutyl ester, hydroxybutyl acrylate, 1,4-butanediol monoacrylate, 2-propenoic acid, 4-hydroxybutyl ester, unii-4o2a4het1x, 4o2a4het1x, hydroxybutylacrylate, 4-hydroxybutylacrylate, 4-hydroxy-butyl acrylate
PubChem CID 75588
IUPAC Name 4-hydroxybutyl prop-2-enoate
SMILES C=CC(=O)OCCCCO

Specifications

Color Yellow
Boiling Point 95°C
Quantity 500 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hydroxybutyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0958
TSCA Yes