missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybutyl acrylate, 95%, stab. with ca 500ppm 4-methoxyphenol

Catalog No. p-7045542
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1109706 5 g
AAL1109714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1109706 Supplier Thermo Scientific Chemicals Supplier No. L1109706

CAS: 2478-10-6 | C7H16ClNO2S | 213.72 g/mol

4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2478-10-6
MDL Number MFCD00010261

Specifications

Melting Point -112°C
Density 1.041
Boiling Point ∼230°C
Flash Point >110°C (230°F)
Molecular Formula C7H12O3
Refractive Index 1.452
Quantity 5 g
UN Number UN3082
Beilstein 1756545
Synonym 4-hydroxybutyl acrylate, acrylic acid 4-hydroxybutyl ester, hydroxybutyl acrylate, 1,4-butanediol monoacrylate, 2-propenoic acid, 4-hydroxybutyl ester, unii-4o2a4het1x, 4o2a4het1x, hydroxybutylacrylate, 4-hydroxybutylacrylate, 4-hydroxy-butyl acrylate
Solubility Information Fully miscible in water.
InChI Key NDWUBGAGUCISDV-UHFFFAOYSA-N
SMILES C=CC(=O)OCCCCO
IUPAC Name 4-hydroxybutyl prop-2-enoate
Molecular Weight (g/mol) 144.17
PubChem CID 75588
Formula Weight 144.17
Percent Purity 95%
Chemical Name or Material 4-Hydroxybutyl acrylate, stabilized with ≈500ppm 4-methoxyphenol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 219-606-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only