missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic acid, sodium salt, 98%

Catalog No. AC391520250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC391520250 Supplier Thermo Scientific Chemicals Supplier No. 391520250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114-63-6
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
MDL Number MFCD00016530
InChI Key ZLVSYODPTJZFMK-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1
PubChem CID 16219477
IUPAC Name sodium;4-hydroxybenzoate
SMILES [Na+].OC1=CC=C(C=C1)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4COONa
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 160.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzoic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-051-1

RUO – Research Use Only