missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonic Acid Hydrate 85.0+%, TCI America™

Catalog No. p-7134425
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
P010425G 25 g
P0104500G 500 g
2 options

Catalog No. P010425G

Supplier: TCI America P010425G

Only null left

Chemical Identifiers

CAS 98-67-9
Molecular Formula C6H6O4S
Molecular Weight (g/mol) 174.17
MDL Number MFCD00007506
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-N
Synonym p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid
PubChem CID 4765
ChEBI CHEBI:32354
IUPAC Name 4-hydroxybenzene-1-sulfonic acid
SMILES OC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Quantity 25 g
UN Number 1759
Formula Weight 174.17
Percent Purity ≥85.0% (T)
Physical Form Crystals
Chemical Name or Material 4-Hydroxybenzenesulfonic Acid Hydrate
EINECSNumber (3)-1956
RTECSNumber DB6970000
TSCA Yes