missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzamidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7131082
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15101G 1 g
H15105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15101G Supplier TCI America Supplier No. H15101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38148-63-9
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00136405
InChI Key OADOZRQXRQAJDT-UHFFFAOYSA-N
Synonym 4-hydroxybenzamidine hydrochloride, 4-amidinophenol hydrochloride, 4-hydroxy-benzamidine hcl, p-hydroxybenzamidine hydrochloride, 4-hydroxybenzimidamide hydrochloride, 4-hydroxybenzenecarboximidamide hydrochloride, 4-hydroxybenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-hydroxy-, monohydrochloride, benzamidine, p-hydroxy-, monohydrochloride, 4-hydroxybenzenecarboximidamide monohydrochloride
PubChem CID 5493520
IUPAC Name hydrogen 4-(diaminomethylidene)cyclohexa-2,5-dien-1-one chloride
SMILES [H+].[Cl-].NC(N)=C1C=CC(=O)C=C1

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 1 g
Formula Weight 172.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzamidine Hydrochloride
RTECSNumber CV6262000
TSCA No