missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzamidine hydrochloride, 95%

Catalog No. p-7030727
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3156603 1 g
AAH3156609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3156603 Supplier Thermo Scientific Chemicals Supplier No. H3156603
Only null left

CAS: 38148-63-9 | C7H9ClN2O | 172.61 g/mol

4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 38148-63-9
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00136405
InChI Key OADOZRQXRQAJDT-UHFFFAOYSA-N
Synonym 4-hydroxybenzamidine hydrochloride, 4-amidinophenol hydrochloride, 4-hydroxy-benzamidine hcl, p-hydroxybenzamidine hydrochloride, 4-hydroxybenzimidamide hydrochloride, 4-hydroxybenzenecarboximidamide hydrochloride, 4-hydroxybenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-hydroxy-, monohydrochloride, benzamidine, p-hydroxy-, monohydrochloride, 4-hydroxybenzenecarboximidamide monohydrochloride
PubChem CID 5493520
IUPAC Name 4-(diaminomethylidene)cyclohexa-2,5-dien-1-one;hydrochloride
SMILES [H+].[Cl-].NC(N)=C1C=CC(=O)C=C1

Specifications

Melting Point 221°C to 224°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.61
Percent Purity 95%
Chemical Name or Material 4-Hydroxybenzamidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber CV6262000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only