missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzamide 98.0+%, TCI America™
$63.38 - $201.32
Chemical Identifiers
| CAS | 619-57-8 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.14 |
| MDL Number | MFCD00007997 |
| InChI Key | QXSAKPUBHTZHKW-UHFFFAOYSA-N |
| Synonym | benzamide, 4-hydroxy, 4-hydroxy-benzamide, p-hydroxybenzamide, unii-9ou5yd093j, 4-hydroxy benzamide, 1,3,4,9-tetrahydro-2-hydroxybenzoyl-9-4-hydroxyphenyl methyl-6-methoxy-2h-pyrido 3,4-b indole, 4-oxidanylbenzamide, acmc-1apvc, para-hydroxybenzoic amide |
| PubChem CID | 65052 |
| IUPAC Name | 4-hydroxybenzamide |
| SMILES | NC(=O)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 619-57-8 | |
| 137.14 | |
| QXSAKPUBHTZHKW-UHFFFAOYSA-N | |
| 65052 | |
| NC(=O)C1=CC=C(O)C=C1 |
| C7H7NO2 | |
| MFCD00007997 | |
| benzamide, 4-hydroxy, 4-hydroxy-benzamide, p-hydroxybenzamide, unii-9ou5yd093j, 4-hydroxy benzamide, 1,3,4,9-tetrahydro-2-hydroxybenzoyl-9-4-hydroxyphenyl methyl-6-methoxy-2h-pyrido 3,4-b indole, 4-oxidanylbenzamide, acmc-1apvc, para-hydroxybenzoic amide | |
| 4-hydroxybenzamide |
Specifications
| 619-57-8 | |
| C7H7NO2 | |
| 5 g | |
| QXSAKPUBHTZHKW-UHFFFAOYSA-N | |
| 4-hydroxybenzamide | |
| 65052 | |
| ≥98.0% (N) | |
| 4-Hydroxybenzamide |
| 161°C | |
| MFCD00007997 | |
| benzamide, 4-hydroxy, 4-hydroxy-benzamide, p-hydroxybenzamide, unii-9ou5yd093j, 4-hydroxy benzamide, 1,3,4,9-tetrahydro-2-hydroxybenzoyl-9-4-hydroxyphenyl methyl-6-methoxy-2h-pyrido 3,4-b indole, 4-oxidanylbenzamide, acmc-1apvc, para-hydroxybenzoic amide | |
| NC(=O)C1=CC=C(O)C=C1 | |
| 137.14 | |
| 137.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No